2-Chloro-N,N-dimethylaniline 97%

Supplier: Ambeed
Warning

A196163-1G A196163-25G
77139-244EA 10.61 USD
77139-244 77685-489
2-Chloro-N,N-dimethylaniline 97%
2-Chloro-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MDL Number: MFCD03452792
CAS Number: 698-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR