(6AR,8R,9R,9aS)-8-(6-amino-9H-purin-9-yl)-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-9-ol 97%
Supplier: Ambeed
|
Warning
|
A194444-1G
A194444-250MG
A194444-5G
77139-198EA
64.88
USD
77139-198
77139-200
77139-202
(6AR,8R,9R,9aS)-8-(6-amino-9H-purin-9-yl)-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-9-ol 97%
(6AR,8R,9R,9aS)-8-(6-amino-9H-purin-9-yl)-2,2,4,4-tetraisopropyltetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-9-ol
Precaución: for Research Use Only
Formula:
C₂₂H₃₉N₅O₅Si₂ |
MDL Number:
MFCD00012186 CAS Number: 69304-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...