4-Bromo-3-chloro-2-methylaniline 96%

Supplier: Ambeed
Warning

A194158-1G A194158-25G A194158-5G
77139-180EA 7.43 USD
77139-180 77139-182 77139-184
4-Bromo-3-chloro-2-methylaniline 96%
4-Bromo-3-chloro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.4942 g/mol
MDL Number: MFCD07369999
CAS Number: 627531-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR