3-Chloro-2-hydrazinopyridine 97%

Supplier: Ambeed
Danger

A193773-10G A193773-25G
77139-158EA 13.8 USD
77139-158 77615-640
3-Chloro-2-hydrazinopyridine 97%
3-Chloro-2-hydrazinopyridine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MDL Number: MFCD01936479
CAS Number: 22841-92-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR