N,N'-(Cyclohexane-1,4-diyl)bis(2-(4-chlorophenoxy)acetamide) 97%

Supplier: Ambeed
Warning

A193697-10MG A193697-25MG A193697-50MG A193697-5MG
77139-148EA 127.72 USD
77139-148 77139-150 77139-152 77139-154 77139-156
N,N'-(Cyclohexane-1,4-diyl)bis(2-(4-chlorophenoxy)acetamide) 97%
N,N'-(Cyclohexane-1,4-diyl)bis(2-(4-chlorophenoxy)acetamide)

Precaución: for Research Use Only

Formula: C₂₂H₂₄Cl₂N₂O₄
MW: 451.34 g/mol
MDL Number: MFCD27952932
CAS Number: 548470-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR