(4-(Piperidin-1-ylsulfonyl)-2-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A193585-100MG A193585-1G A193585-250MG
77139-136EA 37.14 USD
77139-136 77139-138 77139-140
(4-(Piperidin-1-ylsulfonyl)-2-(trifluoromethyl)phenyl)boronic acid 97%
(4-(Piperidin-1-ylsulfonyl)-2-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₃NO₄S
MW: 337.12 g/mol
MDL Number: MFCD18783153
CAS Number: 2096339-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR