1-(2-Bromo-3,4-difluorophenyl)ethanone 97%

Supplier: Ambeed
Danger

A193297-100MG A193297-250MG A193297-5G
77139-126EA 50.93 USD
77139-126 77139-128 77139-130
1-(2-Bromo-3,4-difluorophenyl)ethanone 97%
1-(2-Bromo-3,4-difluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
MDL Number: MFCD26687998
CAS Number: 936846-32-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR