2,5-Dichlorobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (2,5-Dichlorophenyl)methanol

A193079-1G A193079-250MG A193079-5G
77139-120EA 9.55 USD
77139-120 77139-122 77139-124
2,5-Dichlorobenzyl alcohol 97%
2,5-Dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00004607
CAS Number: 34145-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR