(Methoxymethyl)triphenylphosphonium bromide 95%

Supplier: Ambeed
Warning

A192754-5G A192754-25G
77139-114EA 19.57 USD
77139-114 77615-630
(Methoxymethyl)triphenylphosphonium bromide 95%
(Methoxymethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₀H₂₀BrOP
MDL Number: MFCD00075442
CAS Number: 33670-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR