(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97%

Supplier: Ambeed
Warning

A192292-100MG A192292-1G A192292-250MG A192292-25G A192292-5G
77139-090EA 6.18 USD
77139-090 77139-092 77139-094 77139-096 77139-098
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97%
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₂
MDL Number: MFCD00040565
CAS Number: 109838-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR