(S)-(-)-4-Fluoro-α-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(4-Fluorophenyl)ethanol, (1S)-1-(4-Fluorophenyl)ethan-1-ol

A191680-100MG A191680-1G A191680-250MG A191680-5G
77139-050EA 22.65 USD
77139-050 77139-052 77139-054 77139-056
(S)-(-)-4-Fluoro-α-methylbenzyl alcohol 97%
(S)-(-)-4-Fluoro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD03092998
CAS Number: 101219-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR