Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: (2-Dicyclohexylphosphino-2',4',6'-triisopropyl-1,1'-biphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(ii) methanesulphonate

A190330-10G A190330-1G A190330-250MG A190330-25G A190330-5G
77139-030EA 214.24 USD
77139-030 77139-032 77139-034 77139-036 77139-038
Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₆H₆₂NO₃PPdS
MDL Number: MFCD22417234
CAS Number: 1445085-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR