N-(2-Phenoxyphenyl)methanesulfonamide 97%

Supplier: Ambeed
Warning

A190077-100G A190077-10G A190077-1G A190077-250MG A190077-25G
77139-014EA 359.65 USD
77139-014 77139-016 77139-018 77139-020 77139-022
N-(2-Phenoxyphenyl)methanesulfonamide 97%
N-(2-Phenoxyphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃S
MW: 263.31 g/mol
MDL Number: MFCD03389778
CAS Number: 51765-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR