(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one 95%

Supplier: Ambeed
Warning

Synonyms: (3aR,6aR)-2,2-Dimethyl-3aH-cyclopenta[d][1,3]dioxol-4(6aH)-one

A189045-100MG A189045-250MG A189045-25G A189045-5G A189045-1G
77139-004EA 6.37 USD
77139-004 77139-006 77139-008 77139-010 77615-600
(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one 95%
(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD08166479
CAS Number: 115509-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR