3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dichloro-1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, 3,6-Dichlorotrimellitic anhydride

A187969-100MG A187969-1G A187969-250MG A187969-25G A187969-5G A187969-50MG
77138-988EA 65.93 USD
77138-988 77138-990 77138-992 77615-592 77615-594 77746-207
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride 97%
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride

Precaución: for Research Use Only

Formula: C₉H₂Cl₂O₅
MW: 261.0152 g/mol
MDL Number: MFCD28962732
CAS Number: 81742-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR