(-)-Epigallocatechin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol, 3,5,7,3',4',5'-Hexahydroxylflavan
A187930-1G
A187930-250MG
A187930-5G
A187930-5MG
A187930-1MG
77138-980EA
40.17
USD
77138-980
77138-982
77138-984
77138-986
77685-443
(-)-Epigallocatechin 98%
(-)-Epigallocatechin
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₇ MW: 306.27 g/mol |
MDL Number:
MFCD00075939 CAS Number: 970-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...