2-[(4-Methoxybenzoyl)amino]acetic acid 97%

Supplier: Ambeed
Warning

A187302-10G A187302-1G A187302-5G
77138-946EA 180.24 USD
77138-946 77138-948 77138-950
2-[(4-Methoxybenzoyl)amino]acetic acid 97%
2-[(4-Methoxybenzoyl)amino]acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00181815
CAS Number: 13214-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR