(S)-(-)-Menthyltoluene-4-sulfinate 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulphinate, (S)-((1R,2S,5R)-2-isopropyl-5-methylcyclohexyl) 4-methylbenzenesulphinate

A186814-1G A186814-250MG A186814-5G
77138-936EA 13.8 USD
77138-936 77138-938 77138-940
(S)-(-)-Menthyltoluene-4-sulfinate 98%
(S)-(-)-Menthyltoluene-4-sulphinate

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₂S
MW: 294.4521 g/mol
MDL Number: MFCD00010192
CAS Number: 1517-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR