Chidamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Amino-5-fluorophenyl)-4-((3-(pyridin-3-yl)acrylamido)methyl)benzamide, HDAC-IN-7

A186722-10MG A186722-1MG A186722-5MG
77138-930EA 292.81 USD
77138-930 77138-932 77138-934
Chidamide 98%
Chidamide

Precaución: for Research Use Only

Formula: C₂₂H₁₉FN₄O₂
MW: 390.4103 g/mol
MDL Number: MFCD18382123
CAS Number: 743420-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR