Benzyl (6-oxohexyl)carbamate 97%

Supplier: Ambeed
Warning

A186294-1G A186294-250MG A186294-5G
77138-916EA 77.25 USD
77138-916 77138-918 77138-920
Benzyl (6-oxohexyl)carbamate 97%
Benzyl (6-oxohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.30 g/mol
MDL Number: MFCD12031811
CAS Number: 98648-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR