(4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate 97%
Supplier: Ambeed
|
Warning
|
A186222-1G
A186222-250MG
A186222-25G
A186222-5G
77138-908EA
14.42
USD
77138-908
77138-910
77138-912
77138-914
(4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate 97%
(4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)methyl pivalate
Precaución: for Research Use Only
Formula:
C₁₅H₁₇N₅O₂ MW: 299.33 g/mol |
MDL Number:
MFCD28975987 CAS Number: 1146629-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...