1-(2-(Di(adamantan-1-yl)phosphino)phenyl)piperidine 98%

Supplier: Ambeed
Warning

A185914-100MG A185914-1G A185914-250MG
77138-896EA 60.48 USD
77138-896 77138-898 77138-900
1-(2-(Di(adamantan-1-yl)phosphino)phenyl)piperidine 98%
1-(2-(Di(adamantan-1-yl)phosphino)phenyl)piperidine

Precaución: for Research Use Only

Formula: C₃₁H₄₄NP
MW: 461.66 g/mol
MDL Number: MFCD28053539
CAS Number: 1237588-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR