2-Nitro-4-thiocyanatoaniline 97%

Supplier: Ambeed
Warning

A185683-10G A185683-1G A185683-250MG A185683-25G A185683-5G
77138-882EA 31.94 USD
77138-882 77138-884 77138-886 77138-888 77138-890
2-Nitro-4-thiocyanatoaniline 97%
2-Nitro-4-thiocyanatoaniline

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂S
MDL Number: MFCD00463878
CAS Number: 54029-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR