2-Amino-6-fluorobenzamide 98%

Supplier: Ambeed
Warning

A185221-1G A185221-25G A185221-5G
77138-860EA 10.31 USD
77138-860 77138-862 77138-864
2-Amino-6-fluorobenzamide 98%
2-Amino-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇FN₂O
MDL Number: MFCD00816881
CAS Number: 115643-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR