6-Bromo-7-chloroquinazoline-2,4-diol 97%

Supplier: Ambeed
Warning

A185202-100MG A185202-1G A185202-250MG
77138-854EA 35.02 USD
77138-854 77138-856 77138-858
6-Bromo-7-chloroquinazoline-2,4-diol 97%
6-Bromo-7-chloroquinazoline-2,4-diol

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂O₂
MDL Number: MFCD29058761
CAS Number: 1166378-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR