2-Bromo-3,5-difluorobenzaldehyde 97%

Supplier: Ambeed
Warning

A185012-25G A185012-10G A185012-1G A185012-5G
77138-852EA 446.64 USD
77138-852 77746-194 77746-195 77746-196
2-Bromo-3,5-difluorobenzaldehyde 97%
2-Bromo-3,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
MDL Number: MFCD24387232
CAS Number: 1232407-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR