3-Chloro-1,2-benzisoxazole 97%

Supplier: Ambeed
Warning

A184444-1G A184444-25G A184444-5G
77138-834EA 54.11 USD
77138-834 77138-836 77138-838
3-Chloro-1,2-benzisoxazole 97%
3-Chloro-1,2-benzisoxazole

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MW: 153.5658 g/mol
MDL Number: MFCD03407314
CAS Number: 16263-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR