2,4,5-Tribromotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2,4,5-tribromobenzene, 1,2,4-Tribromo-5-methylbenzene

A184017-250MG A184017-5G
77138-830EA 5.31 USD
77138-830 77138-832
2,4,5-Tribromotoluene 98%
2,4,5-Tribromotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₃
MW: 328.83 g/mol
MDL Number: MFCD00052984
CAS Number: 3278-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR