4-Nitrophenyl bromoacetate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenyl 2-bromoacetate

A183977-1G A183977-250MG A183977-5G A183977-10G
77138-824EA 38.2 USD
77138-824 77138-826 77138-828 77685-430
4-Nitrophenyl bromoacetate 95%
4-Nitrophenyl bromoacetate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MW: 260.0415 g/mol
MDL Number: MFCD00051840
CAS Number: 19199-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR