1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(1,4-Dioxaspiro[4.5]decan-8-yl)ethanone

A183974-100MG A183974-1G A183974-250MG
77138-818EA 31.83 USD
77138-818 77138-820 77138-822
1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone 95%
1-(1,4-Dioxaspiro[4.5]dec-8-yl)-ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₃
MW: 184.232 g/mol
MDL Number: MFCD20257858
CAS Number: 35477-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR