(S)-tert-Butyl 6-(aminomethyl)-2,2-dimethylmorpholine-4-carboxylate oxalate 97%

Supplier: Ambeed
Warning

A183496-1G A183496-250MG
77138-808EA 206 USD
77138-808 77138-810
(S)-tert-Butyl 6-(aminomethyl)-2,2-dimethylmorpholine-4-carboxylate oxalate 97%
(S)-tert-Butyl 6-(aminomethyl)-2,2-dimethylmorpholine-4-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₂O₇
MW: 334.36 g/mol
MDL Number: MFCD28291834
CAS Number: 1951425-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR