9,9-Dimethylfluorene-2,7-diboronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(9,9-Dimethyl-9H-fluorene-2,7-diyl)diboronic acid
A183414-1G
A183414-250MG
A183414-5G
77138-794EA
20.6
USD
77138-794
77138-796
77138-798
9,9-Dimethylfluorene-2,7-diboronic acid 95%
9,9-Dimethylfluorene-2,7-diboronic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₆B₂O₄ MW: 281.907 g/mol |
MDL Number:
MFCD08704228 CAS Number: 866100-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...