Ketoconazole 98%

Supplier: Ambeed
Danger

Synonyms: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine

A183059-10G A183059-1G A183059-25G A183059-5G
77138-788EA 11.67 USD
77138-788 77138-790 77138-792 77685-425
Ketoconazole 98%
Ketoconazole

Precaución: for Research Use Only

Formula: C₂₆H₂₈Cl₂N₄O₄
MW: 531.43 g/mol
MDL Number: MFCD00058579
CAS Number: 65277-42-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR