Amino-PEG10-Amine 95%

Supplier: Ambeed
Warning

Synonyms: 3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diamine

A181623-100MG A181623-250MG
77138-752EA 135.8 USD
77138-752 77685-421
Amino-PEG10-Amine 95%
Amino-PEG10-Amine

Precaución: for Research Use Only

Formula: C₂₂H₄₈N₂O₁₀
MW: 500.624 g/mol
MDL Number: MFCD28122962
CAS Number: 928292-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR