6-Methoxy-2-pyridinemethanamine 97%

Supplier: Ambeed
Warning

A181512-1G A181512-5G
77138-736EA 59.74 USD
77138-736 77550-170
6-Methoxy-2-pyridinemethanamine 97%
6-Methoxy-2-pyridinemethanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.16 g/mol
MDL Number: MFCD11617139
CAS Number: 194658-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR