Potassium (4-((tert-butoxycarbonyl)amino)phenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

A181018-1G A181018-250MG A181018-5G
77138-712EA 167.89 USD
77138-712 77138-714 77138-716
Potassium (4-((tert-butoxycarbonyl)amino)phenyl)trifluoroborate 97%
Potassium (4-((tert-butoxycarbonyl)amino)phenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BF₃KNO₂
MDL Number: MFCD26131167
CAS Number: 2244495-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR