(R)-(+)-4-Chloro-α-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Chloro-alpha-methylbenzyl Alcohol, (R)-1-(4-Chlorophenyl)ethanol

A180676-100MG A180676-1G A180676-250MG
77138-698EA 9.27 USD
77138-698 77138-700 77138-702
(R)-(+)-4-Chloro-α-methylbenzyl alcohol 95%
(R)-(+)-4-Chloro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD06797295
CAS Number: 75968-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR