6-(tert-Butyl)-1H-indole 95%

Supplier: Ambeed
Warning

A179857-100MG A179857-1G A179857-250MG
77138-672EA 21.63 USD
77138-672 77138-674 77138-676
6-(tert-Butyl)-1H-indole 95%
6-(tert-Butyl)-1H-indole

Precaución: for Research Use Only

Formula: C₁₂H₁₅N
MDL Number: MFCD07367958
CAS Number: 887581-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR