Benzyl butyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%

Supplier: Ambeed
Warning

A179564-1G A179564-250MG A179564-5G
77138-648EA 32.96 USD
77138-648 77138-650 77138-652
Benzyl butyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%
Benzyl butyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₄H₃₂BNO₄
MW: 409.33 g/mol
MDL Number: MFCD21609689
CAS Number: 2096994-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR