Cyclohexane-1,2-diyldimethanol 97%

Supplier: Ambeed
Warning

A178866-1G A178866-250MG
77138-636EA 49.43 USD
77138-636 77138-638
Cyclohexane-1,2-diyldimethanol 97%
Cyclohexane-1,2-diyldimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD19300921
CAS Number: 3971-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR