Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A178446-100MG A178446-10G A178446-1G A178446-250MG
77138-620EA 17.52 USD
77138-620 77138-622 77138-624 77138-626
Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate 97%
Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MDL Number: MFCD11520455
CAS Number: 15448-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR