Methyl 2,4-dichloro-6-fluorobenzoate 98%

Supplier: Ambeed
Warning

A178155-1G A178155-250MG
77138-616EA 113.52 USD
77138-616 77138-618
Methyl 2,4-dichloro-6-fluorobenzoate 98%
Methyl 2,4-dichloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO₂
MW: 223.02 g/mol
MDL Number: MFCD22574195
CAS Number: 1398504-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR