(1S,2S)-Fmoc-Acpc 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1S,2S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopentanecarboxylic acid
A178090-250MG
A178090-5G
77138-614EA
87
USD
77138-614
77550-142
(1S,2S)-Fmoc-Acpc 95%
(1S,2S)-Fmoc-Acpc
Precaución: for Research Use Only
Formula:
C₂₁H₂₁NO₄ MW: 351.41 g/mol |
MDL Number:
MFCD04112694 CAS Number: 359586-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...