(1R)-1-(Trifluoromethyl)ethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2R)-2-Amino-1,1,1-trifluoropropane hydrochloride, (R)-1,1,1-Trifluoropropan-2-amine hydrochloride, (2R)-1,1,1-Trifluoropropan-2-amine hydrochloride

A177675-10G A177675-1G A177675-250MG A177675-25G A177675-5G
77138-596EA 176.11 USD
77138-596 77138-598 77138-600 77138-602 77138-604
(1R)-1-(Trifluoromethyl)ethylamine hydrochloride 95%
(1R)-1-(Trifluoromethyl)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₆F₃N·ClH
MW: 149.543 g/mol
MDL Number: MFCD07779095
CAS Number: 177469-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR