(R)-3-(4-Bromo-3-fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one 97%

Supplier: Ambeed
Warning

A177584-100G A177584-10G A177584-1G A177584-250MG
77138-588EA 618.51 USD
77138-588 77138-590 77138-592 77138-594
(R)-3-(4-Bromo-3-fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one 97%
(R)-3-(4-Bromo-3-fluorophenyl)-5-(hydroxymethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrFNO₃
MW: 290.09 g/mol
MDL Number: MFCD08166353
CAS Number: 444335-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR