3-Formylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid

A176705-100G A176705-10G A176705-25G A176705-500G A176705-5G
77138-568EA 73.13 USD
77138-568 77138-570 77138-572 77138-574 77138-576
3-Formylphenylboronic acid 97%
3-Formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₃
MDL Number: MFCD00161356
CAS Number: 87199-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR