(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%

Supplier: Ambeed
Warning

A176135-100MG A176135-1G A176135-250MG A176135-25G A176135-5G
77138-556EA 5.15 USD
77138-556 77138-558 77138-560 77138-562 77138-564
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₉FeN
MDL Number: MFCD00066273
CAS Number: 31886-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR