2-(2-(tert-Butyl)phenoxy)pyridin-3-amine 97%

Supplier: Ambeed
Warning

A176100-100MG A176100-1G
77138-552EA 20.16 USD
77138-552 77138-554
2-(2-(tert-Butyl)phenoxy)pyridin-3-amine 97%
2-(2-(tert-Butyl)phenoxy)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O
MW: 242.32 g/mol
MDL Number: MFCD11156638
CAS Number: 861673-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR