1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol ≥99%

Supplier: Ambeed
Warning

Synonyms: 1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(phenylamino)propan-2-ol, P7C3

A176049-100MG A176049-10MG A176049-1MG A176049-50MG A176049-5MG
77138-542EA 30.77 USD
77138-542 77138-544 77138-546 77138-548 77138-550
1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol ≥99%
1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol

Precaución: for Research Use Only

Formula: C₂₁H₁₈Br₂N₂O
MW: 474.21 g/mol
MDL Number: MFCD00572918
CAS Number: 301353-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR