(3aR,6S)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 98%

Supplier: Ambeed
Warning

A175160-1G A175160-25G A175160-5G
77138-524EA 13.8 USD
77138-524 77138-526 77138-528
(3aR,6S)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 98%
(3aR,6S)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.3 g/mol
MDL Number: MFCD00064576
CAS Number: 107869-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR